BAF370840008
SP Bel-Art® Round Microcentrifuge Floating Bubble Rack with Hold-Down Disk
to hold, 8 x 1.5ml Tubes
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
feature
fits beaker size 400 mL
to hold 8 x 1.5ml Tubes
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F37084-0008
O.D.
2⁵/₈ in. (66 mm)
66 mm
hole diam.
0.4375 in. (10.8 mm)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Specifically designed for incubating 1.5 ml microcentrifuge tubes, this rack includes a hold-down disk which maintains a constant pressure on the tube caps and prevents them from opening during internal air expansion.
Features and Benefits
- The polypropylene disk with threaded handle screws into the foam-filled TPX® polymethylpentene plastic rack and holds tubes firmly in place
- Rack floats while microcentrifuge tubes remain immersed at a constant depth (3/4")
- Leg length: 19mm Steam autoclavable at 121ºC (250ºF)
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
TPX is a registered trademark of Mitsui Petrochemical Industries, Ltd.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service