Skip to Content
Merck
All Photos(1)

Documents

W439400

Sigma-Aldrich

2,4,5-Trimethyloxazole

99%, FG

Synonym(s):

Trimethyloxazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9NO
CAS Number:
Molecular Weight:
111.14
FEMA Number:
4394
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.169
NACRES:
NA.21

biological source

synthetic

grade

FG

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

99%

refractive index

n20/D 1.442 (lit.)

bp

133-134 °C/760 mmHg (lit.)

density

0.957 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; nutty; sweet

SMILES string

Cc1nc(C)c(C)o1

InChI

1S/C6H9NO/c1-4-5(2)8-6(3)7-4/h1-3H3

InChI key

ZRLDBDZSLLGDOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4,5-Trimethyloxazole is an important heterocyclic flavor compound that generally occurs in coffee and cocoa. It is reported to be present in meat and roasted peanuts due to its formation by the reaction between 2,3-butanedione and alanine or cysteine due to Strecker′s degradation.

Application

Possible applications: Chocolate, Dutch cocoa, coffee, various savory meaty flavors, malt and toasted bread.

Biochem/physiol Actions

Odor at 1.0%: Slight pyridine-like, roasted vegetative, toasted Dutched cocoa background with a slight nutty nuance.
Taste at 1ppm: Sweet roasted cocoa, coffee, chocolate, slightly seared savory beef, proteinous with green vegetative nuances.

Other Notes

Natural occurrence: Heated beef, cocoa, cooked pork, French fries, autolyzed yeast, cooked egg and fried chicken.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Formation of oxazolines and oxazoles in Strecker degradation of DL-alanine and L-cysteine with 2, 3-butanedione.
Ho CT & Hartman GJ.
Journal of Agricultural and Food Chemistry, 30(4), 793-794 (1982)
Oxazoles and oxazolines in foods.
Maga JA.
Journal of Agricultural and Food Chemistry, 26(5), 1049-1050 (1978)
Gerard Mosciano
Perfumer & Flavorist, 34 (1), 58-59 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service