Skip to Content
Merck
All Photos(2)

Key Documents

B45712

Sigma-Aldrich

2,2-Bis(4-hydroxy-3-methylphenyl)propane

97%

Synonym(s):

4,4′-Isopropylidenedi-o-cresol

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€135.00

€135.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€135.00

About This Item

Linear Formula:
(CH3)2C[C6H3(CH3)OH]2
CAS Number:
Molecular Weight:
256.34
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€135.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

bp

238-240 °C/12 mmHg (lit.)

mp

138-140 °C (lit.)

SMILES string

Cc1cc(ccc1O)C(C)(C)c2ccc(O)c(C)c2

InChI

1S/C17H20O2/c1-11-9-13(5-7-15(11)18)17(3,4)14-6-8-16(19)12(2)10-14/h5-10,18-19H,1-4H3

InChI key

YMTYZTXUZLQUSF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used as a color-developer in thermosensitive recording materials.[1][2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chem. Abstr., 119, 259352g-259352g (1993)
Denshi Shashin Gakkaishi, 32, 18-18 (1993)
Katra Kolšek et al.
Toxicology in vitro : an international journal published in association with BIBRA, 29(1), 8-15 (2014-09-07)
A homeostasis of the glucocorticoid and androgen endocrine system is essential to human health. Their disturbance can lead to various diseases, for example cardiovascular, inflammatory and autoimmune diseases, infertility, cancer. Fifteen widely used industrial chemicals that disrupt endocrine activity were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service