Skip to Content
Merck
All Photos(2)

Key Documents

ALD00400

Sigma-Aldrich

2-Chloro-6-iodobenzoic acid

95%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€96.60

€96.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€96.60

About This Item

Empirical Formula (Hill Notation):
C7H4ClIO2
CAS Number:
Molecular Weight:
282.46
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€96.60


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

solid

reaction suitability

reagent type: ligand
reaction type: C-H Activation

SMILES string

OC(C1=C(Cl)C=CC=C1I)=O

InChI

1S/C7H4ClIO2/c8-4-2-1-3-5(9)6(4)7(10)11/h1-3H,(H,10,11)

InChI key

KNWHWEKCHWFXFZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The following building block was synthesized via a C-H activation method developed by Yu and coworkers. The method was incorporated using the Yu-Wasa Auxiliary (Aldrich 791806) and I2 as the sole oxidant for ortho-iodination from the corresponding benzoic acid derivative.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Chen Wang et al.
Journal of the American Chemical Society, 135(28), 10326-10329 (2013-07-11)
Pd-catalyzed ortho-C-H iodination directed by a weakly coordinating amide auxiliary using I2 as the sole oxidant was developed. This reaction is compatible with a wide range of heterocycles including pyridines, imidazoles, oxazoles, thiazoles, isoxazoles, and pyrazoles.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service