Skip to Content
Merck
All Photos(1)

Key Documents

A96207

Sigma-Aldrich

mono-Methyl azelate

85%, technical grade

Synonym(s):

mono-Methyl nonanedioate, Azelaic acid monomethyl ester, Monomethyl azelate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€383.00

€383.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€383.00

About This Item

Linear Formula:
HOOC(CH2)7COOCH3
CAS Number:
Molecular Weight:
202.25
Beilstein:
1775786
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€383.00


Check Cart for Availability

Request a Bulk Order

grade

technical grade

Quality Level

Assay

85%

form

(Liquid or Viscous Liquid or Semi-Solid to Solid)

refractive index

n20/D 1.446 (lit.)

bp

159-160 °C/3 mmHg (lit.)

mp

22-24 °C (lit.)

density

1.045 g/mL at 20 °C (lit.)

SMILES string

COC(=O)CCCCCCCC(O)=O

InChI

1S/C10H18O4/c1-14-10(13)8-6-4-2-3-5-7-9(11)12/h2-8H2,1H3,(H,11,12)

InChI key

VVWPSAPZUZXYCM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Expansion of the ψ-oxidation system AlkBGTL of Pseudomonas putida GPo1 with AlkJ and AlkH results in exclusive mono-esterified dicarboxylic acid production in E. coli.: This study explores the expansion of the ψ-oxidation system in Pseudomonas putida, leading to the exclusive production of mono-esterified dicarboxylic acids, including mono-methyl azelate. This research highlights the potential for utilizing mono-methyl azelate in the biosynthesis of biodegradable polymers and other eco-friendly applications (van Nuland et al., 2017).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service