Skip to Content
Merck
All Photos(1)

Key Documents

171026

Sigma-Aldrich

Dimethyl azelate

technical grade, 80%

Synonym(s):

Dimethyl nonanedioate, Methyl azelate, Nonanedioic acid dimethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€119.00

€119.00


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
500 ML
€119.00

About This Item

Linear Formula:
CH3OCO(CH2)7COOCH3
CAS Number:
Molecular Weight:
216.27
Beilstein:
1710125
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€119.00


Estimated to ship on16 April 2025


Request a Bulk Order

grade

technical grade

Quality Level

vapor pressure

<1 mmHg ( 20 °C)

Assay

80%

form

liquid

refractive index

n20/D 1.435 (lit.)

bp

156 °C/20 mmHg (lit.)

mp

18 °C

density

1.007 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CCCCCCCC(=O)OC

InChI

1S/C11H20O4/c1-14-10(12)8-6-4-3-5-7-9-11(13)15-2/h3-9H2,1-2H3

InChI key

DRUKNYVQGHETPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Azelaic acid esters do not depigment pigmented guinea pig skin.
M G Wilkerson et al.
Archives of dermatology, 126(2), 252-253 (1990-02-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service