Skip to Content
Merck
All Photos(1)

Key Documents

A70003

Sigma-Aldrich

4-Nitro-1-naphthylamine

97%

Synonym(s):

1-Amino-4-nitronaphthalene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€243.00

€243.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€243.00

About This Item

Linear Formula:
H2NC10H6NO2
CAS Number:
Molecular Weight:
188.18
Beilstein:
2211897
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€243.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

powder

mp

190-193 °C (lit.)

SMILES string

Nc1ccc([N+]([O-])=O)c2ccccc12

InChI

1S/C10H8N2O2/c11-9-5-6-10(12(13)14)8-4-2-1-3-7(8)9/h1-6H,11H2

InChI key

BVPJPRYNQHAOPQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jo-Ann Osei-Twum et al.
Drug development and industrial pharmacy, 41(7), 1130-1136 (2014-06-26)
Amphotericin B (AmB) is a highly efficacious therapeutic for invasive fungal infections and protozoal diseases. Increasing prevalence of these conditions warrants the development of an oral AmB formulation. Efflux transporters, such as the ABCB1 gene product P-glycoprotein, affect the oral

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service