Skip to Content
Merck
All Photos(2)

Key Documents

97233

Sigma-Aldrich

1,3-Di-Z-2-(trifluoromethylsulfonyl)guanidine

≥98.0% (HPLC)

Synonym(s):

1,3-Bis(benzyloxycarbonyl)-2-(trifluoromethylsulfonyl)guanidine, 2-(Trifluoromethylsulfonyl)-1,3-di-Z-guanidine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€1,290.00

€1,290.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€1,290.00

About This Item

Empirical Formula (Hill Notation):
C18H16F3N3O6S
CAS Number:
Molecular Weight:
459.40
Beilstein:
7956609
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

€1,290.00


Estimated to ship on29 May 2025


Request a Bulk Order

Assay

≥98.0% (HPLC)

form

crystals

application(s)

peptide synthesis

SMILES string

FC(F)(F)S(=O)(=O)N=C(NC(=O)OCc1ccccc1)NC(=O)OCc2ccccc2

InChI

1S/C18H16F3N3O6S/c19-18(20,21)31(27,28)24-15(22-16(25)29-11-13-7-3-1-4-8-13)23-17(26)30-12-14-9-5-2-6-10-14/h1-10H,11-12H2,(H2,22,23,24,25,26)

InChI key

GLZHTZVKRCUQRB-UHFFFAOYSA-N

Other Notes

Guanidinylation reagent[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K. Feichtinger et al.
The Journal of Organic Chemistry, 63, 3804-3804 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service