8.07666
Sarcosine
for peptide synthesis
Synonym(s):
Sarcosine, Methylaminoacetic acid, N-Methylglycine
Select a Size
About This Item
Application
- Enhancement of biogenic non-extractable residue formation: Sarcosine′s role is examined in the context of nitrogen-fertilizer addition to soil, where it enhances the formation of biogenic non-extractable residues from glyphosate, with implications for agricultural chemical management (Aslam and Nowak, 2024).
- PolySarcosine as an alternative to PEGylation: Recent advancements highlight polySarcosine as a promising alternative to PEGylation for biomedical applications, underscoring its synthesis and utility in enhancing drug delivery systems (Kabil et al., 2024).
- Sensitive sarcosine detection: A novel ratiometric fluorescent assay employs MOF nanozyme-mediated tandem catalysis guided by a logic circuit for highly sensitive detection of sarcosine, enabling smartphone-based portable sensing applications (Hou et al., 2024).
- Photothermal biosensing for sarcosine quantification: A microfluidic paper-based analytical device integrated with photothermal biosensing offers a new method for the sensitive quantification of sarcosine, facilitating rapid and efficient detection (Khachornsakkul and Leelasattarathkul, 2024).
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service