Skip to Content
Merck
All Photos(1)

Documents

938726

Sigma-Aldrich

ImPy-Dipop-Dipp

new

Synonym(s):

5-(2,6-diisopropoxyphenyl)-2-(2,6-diisopropylphenyl)imidazo[1,5-a]pyridin-2-ium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H39ClN2O2
CAS Number:
Molecular Weight:
507.11
UNSPSC Code:
12352100

form

powder

Quality Level

reaction suitability

reaction type: Cross Couplings

color

white

SMILES string

CC(C1=CC=CC(C(C)C)=C1[N+]2=CN3C(C4=C(OC(C)C)C=CC=C4OC(C)C)=CC=CC3=C2)C.[Cl-]

InChI

1S/C31H39N2O2.ClH/c1-20(2)25-13-10-14-26(21(3)4)31(25)32-18-24-12-9-15-27(33(24)19-32)30-28(34-22(5)6)16-11-17-29(30)35-23(7)8;/h9-23H,1-8H3;1H/q+1;/p-1

InChI key

XMDMIAJKGFBECK-UHFFFAOYSA-M

General description

ImPy-Dipop-Dipp is an NHC precursor developed in the Szostak group. The ligand is useful in a range of catalytic transformations, and is particulary well suited to the following: Suzuki-Miyaura cross-coupling of aryl mesylates and tosylates, Suzuki–Miyaura cross-coupling of nitroarenes (Ar–NO2), Buchwald-Hartwig amination of alkyl sulfoxides

Application

Suzuki-Miyaura cross-coupling of aryl mesylates and tosylates Suzuki–Miyaura cross-coupling of nitroarenes (Ar–NO2) or aryl sulfamates Buchwald-Hartwig amination of alkyl sulfoxides or aryl sulfamates

Features and Benefits

ImPy-Dipop-Dipp is an asymmetric NHC ligand precursor. It forms air-and moisture stable complexes with transition metals and is particulary effective in palladium catalyzed cross-coupling reactions.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service