Skip to Content
Merck
All Photos(1)

Key Documents

715417

Sigma-Aldrich

1,3-Bis(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-2-ylidene

Synonym(s):

1,3-Bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene, SIMes

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€76.90

€76.90


Estimated to ship on12 April 2025


Request a Bulk Order

Select a Size

Change View
250 MG
€76.90

About This Item

Empirical Formula (Hill Notation):
C21H26N2
CAS Number:
Molecular Weight:
306.44
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€76.90


Estimated to ship on12 April 2025


Request a Bulk Order

form

powder

Quality Level

reaction suitability

reagent type: catalyst

mp

79-85 °C

storage temp.

−20°C

SMILES string

Cc1cc(C)c(N2[C]N(CC2)c3c(C)cc(C)cc3C)c(C)c1

InChI

1S/C21H26N2/c1-14-9-16(3)20(17(4)10-14)22-7-8-23(13-22)21-18(5)11-15(2)12-19(21)6/h9-12H,7-8H2,1-6H3

InChI key

LSMWOQFDLBIYPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao Mei et al.
BMC genomics, 16, 336-336 (2015-04-23)
Genetic heritability and expression study have shown that different diabetes traits have common genetic components and pathways. A computationally efficient pathway analysis of GWAS results will benefit post-GWAS study of SNP associations and identification of common genetic pathways from diabetes

Articles

The Hazari group has developed an improved palladium precatalyst scaffold for a wide range of cross-coupling reactions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service