Skip to Content
Merck
All Photos(1)

Key Documents

935387

Sigma-Aldrich

Ready-to-Cast LiMn2O4 (LMO) Slurry for Lithium ion battery

Blade coatable ink slurry for lithium ion battery

Synonym(s):

LMO cathode slurry

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352402
NACRES:
NA.21
Pricing and availability is not currently available.

grade

battery grade

Quality Level

form

slurry

composition

Active material, >36 wt. %
Solid content, 40-60 wt. %

particle size

6-7 μm (APS)

application(s)

battery manufacturing

General description

Our ready-to-cast lithium manganese dioxide (LMO) slurry is a liquid-type dispersion, composed of high-purity spinel-type LMO (D50 particle size 12-18 microns), conductive additives, binder, and organic solvent. The product features a high loading content and excellent stability, ensuring superior performance and uniformity in battery fabrication.

Application

Our ready-to-cast LMO cathode slurry is specifically designed to enable the production of high-performance LMO cathodes for lithium-ion batteries. The slurry can be easily coated onto aluminum foil current collectors using blade-coating or slot-die coating techniques. Due to the precise formulation of our slurry, it is stable and processable, ensuring uniform and defect-free films of silicon anode material. The high active content and excellent stability of our ready-to-cast LMO cathode slurry result in superior battery performance. Researchers can readily use our slurry to optimize their lithium-ion battery fabrication process, resulting in high-performance batteries suitable for a variety of applications.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conveying Advanced Li-ion Battery Materials into Practice The Impact of Electrode Slurry Preparation Skills
Kraytsberg A, et al.
Advanced Energy Materials, 6 (2016)
An Effective Mixing for Lithium Ion Battery Slurries
Liu D, et al.
Advances in Chemical Engineering and Science, 04 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service