Skip to Content
Merck
All Photos(1)

Key Documents

916439

Sigma-Aldrich

Lithium Manganese Oxide spinel (LMO) powder

battery grade

Synonym(s):

LMO, Lithium manganese(III,IV) oxide, NANOMYTE® BE-30

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€224.00

€224.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€224.00

About This Item

Empirical Formula (Hill Notation):
LiMn2O4
CAS Number:
Molecular Weight:
180.81
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.23

€224.00


Check Cart for Availability

Request a Bulk Order

grade

battery grade

Quality Level

description

Charge (First cycle – 4.2V, C/10): 110 mAh/g ± 5%
Discharge (First cycle – 3V, C/10): ≥ 100 mAh/g

form

powder

mol wt

Mw 180.81 g/mol

composition

LiMn2O4

grain size

30-50 nm

avg. part. size

6-7 μm (APS)

density

4.0  g/cm3 (lit.)

application(s)

battery manufacturing

SMILES string

[Mn](=O)=O.[Mn](=O)[O-].[Li+]

InChI key

VLXXBCXTUVRROQ-UHFFFAOYSA-N

Related Categories

General description

Lithium Manganate (LiMn2O4) is a cathode material with a spinel structure, which allows the material to be discharged at high rates. LMO-based batteries are most suited for use in high rate applications.

Application

  • Impact of gadolinium doping into the frustrated antiferromagnetic lithium manganese oxide spinel.: This study explores the effects of gadolinium doping on the properties of lithium manganese oxide spinel, enhancing its application in high-performance batteries (Saini et al., 2023).
  • Oriented LiMn2O4 Particle Fracture from Delithiation-Driven Surface Stress.: This research investigates the fracture mechanisms of LiMn2O4 particles during delithiation, which is crucial for improving the durability and performance of lithium manganese oxide batteries (Warburton et al., 2020).
  • Quantitative analysis of cation mixing and local valence states in LiNixMn2-xO4 using concurrent HARECXS and HARECES measurements.: This paper presents a detailed analysis of cation mixing and valence states in LiNixMn2-xO4, providing insights into the material′s structural and electrochemical properties for battery applications (Yamamoto et al., 2016).
  • Doubling the capacity of lithium manganese oxide spinel by a flexible skinny graphitic layer.: This study demonstrates a method to double the capacity of lithium manganese oxide spinel through the application of a graphitic layer, highlighting significant improvements in battery capacity (Noh et al., 2014).

Legal Information

NANOMYTE is a registered trademark of NEI Corporation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhengxin Zhu et al.
Nano letters, 20(5), 3278-3283 (2020-04-18)
Rechargeable hydrogen gas batteries show promises for the integration of renewable yet intermittent solar and wind electricity into the grid energy storage. Here, we describe a rechargeable, high-rate, and long-life hydrogen gas battery that exploits a nanostructured lithium manganese oxide
Lithium and lithium ion batteries for applications in microelectronic devices
Y Wang, et al.
Journal of Power Sources, 286, 330-345 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service