932876
Tweezer
EMS 2A SA/Dr
Synonym(s):
Gripper, Tweezer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
General description
EMS techware
Application
Safe ergonomic cushion grips (resistivity 10E9 Ohm) for handling ESD sensitive components or small static items. Soft ESD safe cushion grips (resistively 10E8 ohm), these tweezers are ergonomically designed.
OAL: 115 mm. Tips: Flat round.
OAL: 115 mm. Tips: Flat round.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service