Skip to Content
Merck
All Photos(1)

Key Documents

932965

Sigma-Aldrich

Tweezer

Style SM103, Stainless Steel, Epoxy Coated

Synonym(s):

Gripper, Tweezer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NB.35
Pricing and availability is not currently available.

Looking for similar products? Visit Product Comparison Guide

General description

EMS techware

Application

ESD Epoxy Coated Tweezers: This coating provides an ESD Safe coating with surface resistance of 10^5 - 10^6.Non-magnetizable. Typical applications include tweezers for the electronic industry, watch-makers, jewelers and laboratory in moderately aggressive chemical environments. ESD epoxy coated tweezers with handling, positioning at 45° angle. Length: 120mm. ESD epoxy coated tweezers. Cutting tweezers with predominantly angled blade.

OAL: 115 mm Style: Style 14AGW. Material: Carbon Steel.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service