Skip to Content
Merck
All Photos(1)

Key Documents

93093

Sigma-Aldrich

Triphenylphosphine, polymer-bound

100-200 mesh, extent of labeling: ~3 mmol/g triphenylphosphine loading

Synonym(s):

Copolymer of styrene and divinylbenzene, diphenylphosphinated, Diphenylphosphino-polystyrene, Polystyrene crosslinked with divinylbenzene, diphenylphosphinated

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
-C6H4P(C6H5)2
CAS Number:
Molecular Weight:
262.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reaction type: solution phase peptide synthesis
reagent type: ligand

extent of labeling

~3 mmol/g triphenylphosphine loading

matrix

crosslinked with 2% DVB

particle size

100-200 mesh

functional group

phosphine oxide

SMILES string

c1ccc(cc1)P(c2ccccc2)c3ccccc3

InChI

1S/C18H15P/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H

InChI key

RIOQSEWOXXDEQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Polymer-bound triphenylphosphine is a solid-supported reagent[1] that can be used as a better alternative to triphenylphosphine in the Mitsunobu reaction.[2][3]
It can also be used:
  • For the esterification of alkylphosphonic acids using primary alcohols in the presence of iodine and imidazole.[4]
  • To catalyze the conversion of 3-[3-(1,3-dioxolan-2-yl)-1-hydroxypropyl]pyridine to 3-[1-bromo-3-(1,3-dioxolan-2-yl)propyl]pyridine using carbon tetrabromide.[5]
  • To isomerize (Z)-nitro olefins to the (E)-isomers.[6]
  • To prepare polymer-bound ylides which are useful in Wittig reactions.[7]
  • To convert alcohols or carboxylic acids to the corresponding chlorides.[8]
  • In combination with carbon tetrachloride for the coupling N-alkoxycarbonyl α-amino acids and primary amines to form the corresponding amides.[9]

Other Notes

Polymer-bound triphenylphosphine is used for preparing polymer-bound yields which are highly useful for Wittig reactions; removal of the polymer at the end of the reaction removes all the phosphine oxide.[10][11]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Bernard et al.
The Journal of Organic Chemistry, 48, 3164-3164 (1983)
Wittig reagents bound to crosslinked polystyrenes.
Bernard M and Ford W T
The Journal of Organic Chemistry, 48(3), 326-332 (1983)
Multi-step organic synthesis using solid-supported reagents and scavengers: a new paradigm in chemical library generation.
Ley S V, et al.
Journal of the Chemical Society. Perkin Transactions 1, 23, 3815-4195 (2000)
Mild and efficient esterification of alkylphosphonic acids using polymer-bound triphenylphosphine.
Purohit A K, et al.
Tetrahedron Letters, 53(29), 3795-3797 (2012)
The Use of Polymer-Bound Triphenylphosphine in the Stereochemical Inversion of Secondary Alcohols.
White J M, et al.
Combinatorial Chemistry & High Throughput Screening, 3(2), 103-106 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service