Skip to Content
Merck
All Photos(1)

Key Documents

924911

Sigma-Aldrich

25-Hydroxyvitamin-D2-(26,26,26,27,27,27-d6)

≥98 atom % D, ≥95% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€913.00

€913.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 MG
€913.00

About This Item

Empirical Formula (Hill Notation):
C28D6H38O2
CAS Number:
Molecular Weight:
418.68
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.13

€913.00


Check Cart for Availability

Request more information

isotopic purity

≥98 atom % D

Quality Level

Assay

≥95% (CP)

mass shift

M+6

SMILES string

C[C@@]12[C@](CC[C@]2([H])[C@H](C)/C=C/[C@H](C)C(C([2H])([2H])[2H])(C([2H])([2H])[2H])O)([H])/C(CCC1)=C/C=C(C[C@@H](O)CC3)\C3=C

InChI

1S/C28H44O2/c1-19-10-14-24(29)18-23(19)13-12-22-8-7-17-28(6)25(15-16-26(22)28)20(2)9-11-21(3)27(4,5)30/h9,11-13,20-21,24-26,29-30H,1,7-8,10,14-18H2,2-6H3/b11-9+,22-12+,23-13-/t20-,21+,24+,25-,26+,28-/m1/s1/i4D3,5D3

InChI key

KJKIIUAXZGLUND-SGQRFKNGSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - STOT RE 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service