Skip to Content
Merck
All Photos(2)

Key Documents

923230

Sigma-Aldrich

2,4-Spiro-OMeTAD

greener alternative

≥98.5% (HPLC)

Synonym(s):

2,4-Spiro-OMeTAD, N2,N2′,N7,N7′-Tetrakis(2,4-dimethoxyphenyl)-N2,N2′,N7,N7′-tetraphenyl-9,9′-spirobi[fluorene]-2,2′,7,7′-tetraamine

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€339.00
1 G
€531.00

€339.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€339.00
1 G
€531.00

About This Item

Empirical Formula (Hill Notation):
C81H68N4O8
CAS Number:
Molecular Weight:
1225.43
UNSPSC Code:
41116107
NACRES:
NA.23

€339.00


Check Cart for Availability

Request a Bulk Order

description

Band gap 3.2 eV

Quality Level

Assay

≥98.5% (HPLC)

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

solubility

chloroform: soluble

Orbital energy

HOMO -5.24 eV 
LUMO -2.08 eV 

greener alternative category

SMILES string

COC1=CC(OC)=CC=C1N(C2=CC=CC=C2)C3=CC=C4C(C5(C(C=C(N(C6=C(OC)C=C(OC)C=C6)C7=CC=CC=C7)C=C8)=C8C9=C5C=C(N(C%10=CC=C(OC)C=C%10OC)C%11=CC=CC=C%11)C=C9)C%12=C4C=CC(N(C%13=C(OC)C=C(OC)C=C%13)C%14=CC=CC=C%14)=C%12)=C3

InChI key

WOFKCKQGJWGUFS-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to enabling category of greener alternatives, and has been enhanced for energy efficiency. Click here for more information.

Application

High-mobility organic semiconductor hole transport material (HTM) for use in electron transport layer in sensors, OPV and OLED applications. Derivatives of spiro-OMeTAD have been widely studied because of their outstanding performance. It was demonstrated that 2,4-spiro-OMeTAD as HTM in photovoltaic cells shows excelent stability and power conversion efficiency in perovskite solar cells[1]. Zhang et al reported a power conversion efficiency of 17.2% and solar cell filling factor of 70.1% when using 2,4-spiro-OMeTAD.
Spiro-MeOTAD can be used as a hole transport material in perovskite solar cells and solid-state dye-sensitized solar cells (DSSCs) as it helps to efficiently extract and transport positive charges (holes) from the perovskite layer to the electrode, improving the overall device performance and stability. It can be used as a hole injection and transport material in OLEDs, especially in small molecule-based and solution-processed OLEDs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure-performance relationship on the asymmetric methoxy substituents of spiro-OMeTAD for perovskite solar cells.
Zhang M D, et al.
Solar Energy Mat. and Solar Cells, 176, 318-323 (2018)

Articles

Novel biosensing devices, like bio-FET sensors, offer label-free analysis by detecting ionic and biomolecular charges, crucial for diagnostics and biological research.

Explore the eco-friendly potential of organic thin film transistors (OTFTs) for detecting chemical analytes, identifying viruses, and assisting in health diagnostics. This mini-review highlights challenges of achieving sustainability, safety, and biodegradability of each component of an OTFT sensor.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service