Skip to Content
Merck
All Photos(2)

Key Documents

900168

Sigma-Aldrich

Lead(II) iodide

99.999% trace metals basis, perovskite grade

Synonym(s):

Lead diiodide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€149.00

€149.00


Estimated to ship on24 March 2025


Request a Bulk Order

Select a Size

Change View
5 G
€149.00

About This Item

Linear Formula:
PbI2
CAS Number:
Molecular Weight:
461.01
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999% trace metals basis
form:
beads

€149.00


Estimated to ship on24 March 2025


Request a Bulk Order

description

Powder or beads

Quality Level

Assay

99.999% trace metals basis

form

beads

bp

954 °C (lit.)

mp

402 °C (lit.)

density

6.16 g/mL at 25 °C (lit.)

SMILES string

I[PbH2]I

InChI

1S/2HI.Pb/h2*1H;/q;;+2/p-2

InChI key

RQQRAHKHDFPBMC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Lead(II)iodide is a wide bandgap (2.32 eV) semiconductor material. It has uniqueproperties like high resistivity, chemical stability, and a wide range oftemperature applications (−200 °C up to +130 °C). Perovskites with lead as the central cation produce the bestphotovoltaic efficiency. We offer lead iodide, specifically designed forenhanced solar cell performance.

Application

Lead iodide finds application in synthesis of perovskites based photovoltaic materials.[1][2] Our perovskite grade PbI2 can readily be dissolved in DMF to yield 1M solution.
Lead(II) iodide (PbI2) in perovskite grade refers to high-purity lead iodide specifically used for the synthesis and fabrication of perovskite materials, such as formamidinium lead iodide (FAPbI3) or methylammonium lead iodide(MAPbI3), which are commonly studied for perovskite solar cells and other optoelectronic devices. The high purity of lead(II) iodide perovskite grade contributes to the efficiency, stability and reproducibility of perovskite solar cells.

Other Notes

Very air and moisture sensitive. Please handle under argon atmosphere in a glove box.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gratzel M, et al.
Advances in Functional Materials, 25, 6936-6936 (2015)
Samuel D Stranks et al.
Nature nanotechnology, 10(5), 391-402 (2015-05-08)
Metal-halide perovskites are crystalline materials originally developed out of scientific curiosity. Unexpectedly, solar cells incorporating these perovskites are rapidly emerging as serious contenders to rival the leading photovoltaic technologies. Power conversion efficiencies have jumped from 3% to over 20% in

Articles

To achieve net-zero emissions by 2050, renewable power contributions must triple. Photovoltaic stations provide vital utility power, achieved primarily through third- and fourth-generation technology. Promising trends include recycling and revolutionary, ultra-lightweight, flexible, and printable solar cells.

Dr. Perini and Professor Correa-Baena discuss the latest research and effort to obtain higher performance and stability of perovskite materials.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service