Skip to Content
Merck
All Photos(1)

Key Documents

922641

Sigma-Aldrich

Pr-DMQA[BF4]

≥95%

Synonym(s):

Propyl dimethoxyquinacridinium tetrafluoroborate

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€121.00
250 MG
€369.00

€121.00


Estimated to ship on02 April 2025


Request a Bulk Order

Select a Size

Change View
50 MG
€121.00
250 MG
€369.00

About This Item

Empirical Formula (Hill Notation):
C27H29BF4N2O2
CAS Number:
Molecular Weight:
500.34
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

€121.00


Estimated to ship on02 April 2025


Request a Bulk Order

Quality Level

Assay

≥95%

form

powder

reaction suitability

reaction type: Photocatalysis
reagent type: catalyst

storage temp.

−20°C

Application

Pr-DMQA[BF4] is an organic photoredox catalyst for red-light mediated photoreductions and photooxidations.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liangyong Mei et al.
Journal of the American Chemical Society, 142(28), 12056-12061 (2020-07-01)
Red light has the advantages of low energy, less health risks, and high penetration depth through various media. Herein, a helical carbenium ion (N,N'-di-n-propyl-1,13-dimethoxyquinacridinium (

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service