Skip to Content
Merck
All Photos(3)

Key Documents

908177

Sigma-Aldrich

3DPAFIPN

≥95%

Synonym(s):

2,4,6-Tris(diphenylamino)-5-fluoroisophthalonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€119.00
500 MG
€346.00

€119.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€119.00
500 MG
€346.00

About This Item

Empirical Formula (Hill Notation):
C44H30FN5
CAS Number:
Molecular Weight:
647.74
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

€119.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95%

form

powder

reaction suitability

reaction type: Photocatalysis
reagent type: catalyst

mp

307-401 °C

photocatalyst activation

390 nm

SMILES string

Fc1c(c(c(c(c1N(c7ccccc7)c6ccccc6)C#N)N(c5ccccc5)c4ccccc4)C#N)N(c3ccccc3)c2ccccc2

InChI key

GCGRXVMGBHYMDA-UHFFFAOYSA-N

Application

3DPAFIPN is cyanoarene-based donor-acceptor photocatalyst developed by the Zeitler group.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elisabeth Speckmeier et al.
Journal of the American Chemical Society, 140(45), 15353-15365 (2018-10-03)
The targeted choice of specific photocatalysts has been shown to play a critical role for the successful realization of challenging photoredox catalytic transformations. Herein, we demonstrate the successful implementation of a rational design strategy for a series of deliberate structural

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service