Skip to Content
Merck
All Photos(1)

Key Documents

902403

Sigma-Aldrich

3D Printing Hydroxyapatite ink

greener alternative

Synonym(s):

3D printing Hydroxylapatite ink, Calcium phosphate hydroxide, Durapatite, Hydroxyapatite, Hydroxylapatite, Hyperelastic bone

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€494.00

€494.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
€494.00

About This Item

UNSPSC Code:
12352305
NACRES:
NA.23

€494.00


Check Cart for Availability

Request a Bulk Order

form

liquid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

viscosity

20-60 Pa.s

greener alternative category

storage temp.

2-8°C

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". These inks are ideal elements for 3D printed energy storage devices. Click here for more information.

Application

This product can be used in material extrusion 3D printing technique.
This research grade (not for human use) Hydroxyapatite or Hyper elastic Bone 3D-printing ink can be used for printing using a direct extrusion (pneumatic or mechanical) 3D-printer into multi-layered, three-dimensional objects at room temperature. This ink should only be used in a well-ventilated environment. The extruded material rapidly solidifies and allows creation of complex, user-designed forms. Despite being mostly hydroxyapatite, the printed solid material is flexible (may even have limited elastic properties). The solids loading of the as-dried material is 85 vol.% hydroxyapatite, 15 vol.% PLGA. This Hyper elastic Bone ink can be utilized to create three-dimensional structures on their own, or co-3D-printed with other 3D-printing inks to create multi-material structures. The ink may also be mixed with other 3D-printing inks to compound them prior to utilization. Due to the mechanical flexibility of the material after drying, complex 2D and 3D forms can be created. This ink may also be utilized in non-3D-printing applications such as coating (dip-coating or direct painting), thread fabrication, or casting into molds.
Our Hyper elastic Bone 3D-printing ink is comprised of micron sized hydroxyapatite suspended in a mixture of organic solvents containing a dissolved, high molecular weight elastomeric, biocompatible polyester (polylactic-co-glycolic acid;PLGA). Prior to further use, to remove residual solvents after 3D-printing (or other application such as coating, direct painting etc) the composite material should be washed in 70% ethanol and then by water. The 3D-printed hydroxyapatite can be used as a flexible, bioactive, biocompatible ceramic composite or it can also be sintered (1500 °C recommended) in a non-oxidative atmosphere to yield Hyper elastic Bone parts. As sintered, Hyper elastic Bone is highly chemically and thermally stable, and can be used for a wide variety of high temperature applications. It also has numerous hard biological tissue (teeth and bone) applications, and can serve in it as as-printed, washed form as flexible ceramic composite cell scaffold, or in its sintered form, as a rigid ceramic scaffold/implant. In its 3D-printed, flexible form, Hyper elastic Bone has been shown to be highly osteoconductive, and osteogenic, capable of differentiation adult human stem cells into osteoblast-like cells.

Caution

Prior to use mix until the ink becomes homogeneous

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

152.6 °F

Flash Point(C)

67 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service