Skip to Content
Merck
All Photos(1)

Documents

920223

Sigma-Aldrich

3,6-Di-tert-butyl-9-(2,6-dimethylphenyl)-10-(4-(trifluoromethyl)phenyl)acridin-10-ium tetrafluoroborate

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H37BF7N
CAS Number:
Molecular Weight:
627.49
UNSPSC Code:
12352103
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder or crystals

reaction suitability

reaction type: Photocatalysis
reagent type: catalyst

storage temp.

−20°C

Application

3,6-Di-tert-butyl-9-(2,6-dimethylphenyl)-10-(4-(trifluoromethyl)phenyl)acridin-10-ium tetrafluoroborate is a robust acridinium-based photocatalyst reported by Nicewicz and coworkers as an alternative to transition-metal-based photocatalysts with higher chemical stability and attenuated redox potential.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vincent A Pistritto et al.
Journal of the American Chemical Society, 142(40), 17187-17194 (2020-09-29)
Nucleophilic aromatic substitution (SNAr) is a classical reaction with well-known reactivity toward electron-poor fluoroarenes. However, electron-neutral and electron-rich fluoro(hetero)arenes are considerably underrepresented. Herein, we present a method for the nucleophilic defluorination of unactivated fluoroarenes enabled by cation radical-accelerated nucleophilic aromatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service