Skip to Content
Merck
All Photos(2)

Key Documents

909181

Sigma-Aldrich

2,7-Dimethyl-9-mesityl-10-methylacridinium tetrafluoroborate

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€175.00

€175.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€175.00

About This Item

Empirical Formula (Hill Notation):
C25H26BF4N
CAS Number:
Molecular Weight:
427.29
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

€175.00


Check Cart for Availability

Request a Bulk Order

form

powder

reaction suitability

reaction type: Photocatalysis
reagent type: catalyst

SMILES string

[B-](F)(F)(F)F.[n+]1(c2c(c(c4c1ccc(c4)C)c3c(cc(cc3C)C)C)cc(cc2)C)C

InChI key

UCBSHEKJRRAYNP-UHFFFAOYSA-N

Related Categories

Application

Robust acridinium-based photocatalyst reported by Nicewicz and coworkers as an alternative to transition-metal-based photocatalysts with higher chemical stability and attenuated redox potential.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Dale J Wilger et al.
Nature chemistry, 6(8), 720-726 (2014-07-24)
The direct anti-Markovnikov addition of strong Brønsted acids to alkenes remains an unsolved problem in synthetic chemistry. Here, we report an efficient organic photoredox catalyst system for the addition of HCl, HF and also phosphoric and sulfonic acids to alkenes

Related Content

Nicewicz lab focuses on reactive cation radical species for selective reactions, advancing synthesis methodologies for various functional groups.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service