Skip to Content
Merck
All Photos(1)

Key Documents

915173

Sigma-Aldrich

LATP-coated Lithium Manganese Nickel Oxide (LMNO)

powder, battery grade

Synonym(s):

Lithium Aluminum Titanium Phosphate -coated Lithium Manganese Nickel Oxide, NANOMYTE® SP-10C

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€395.00

€395.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€395.00

About This Item

Linear Formula:
LiMn1.5Ni0.5O4 + Li1.4Al0.4Ti1
UNSPSC Code:
26111700
NACRES:
NA.23

€395.00


Check Cart for Availability

Request a Bulk Order

grade

battery grade

Quality Level

description

Minimum Capacity: 110 mAh/g
Nominal Capacity (0.1C): ≥125 mAh/g

form

powder

composition

LiMn1.5Ni0.5O4 coated with 0.1 wt% Li1.3Al0.3Ti1.7(PO4)3

avg. part. size

5-8 μm (APS)

particle size

5-8 μm

grain size

50-70 nm

application(s)

battery manufacturing

Application

LATP coated Lithium manganese nickel oxide is a cathode material that can be charged at high voltage. Due to the high potential, the material has a higher energy density compared to lithium cobalt oxide and lithium iron phosphate.

Legal Information

NANOMYTE is a registered trademark of NEI Corporation

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The lithium-ion battery: State of the art and future perspectives.
Zubi R, et al.
Renewable and Sustainable Energy Reviews, 89, 292-308 (2018)
Li-ion battery materials: present and future.
Nitta N, et al.
Materials Today Communications, 18(5), 252-264 (2015)
The lithium-ion battery: State of the art and future perspectives
G Zubi, et al.
Renewable and Sustainable Energy Reviews, 89, 292-308 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service