Skip to Content
Merck
All Photos(2)

Key Documents

913502

Sigma-Aldrich

Si-OMeTPA

98%

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€268.00

€268.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€268.00

About This Item

Empirical Formula (Hill Notation):
C80H72N4O8Si
Molecular Weight:
1245.54
UNSPSC Code:
12352103
NACRES:
NA.23

€268.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

powder

UV absorption

λ: 221 nm Amax
λ: 313 nm Amax

Application

This is a dopant free, high-performance hole transport material having outstanding performance compared to SpiroMeOTAD.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Severin N Habisreutinger et al.
The journal of physical chemistry letters, 5(23), 4207-4212 (2015-08-19)
Here, we report the use of polymer-wrapped carbon nanotubes as a means to enhance charge extraction through undoped spiro-OMeTAD. With this approach a good solar cell performance is achieved without the implementation of conventional doping methods. We demonstrate that a
Dopant-Free Planar n-i-p Perovskite Solar Cells with Steady-State Efficiencies Exceeding 18%
S N Habisreutinger, et al.
ACS Energy Letters, 2, 622-622 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service