Skip to Content
Merck
All Photos(1)

Key Documents

914545

Sigma-Aldrich

Cesium chloride

AnhydroBeads, 99.999% trace metals basis, (Perovskite grade)

Synonym(s):

Cesium monochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€140.00

€140.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€140.00

About This Item

Linear Formula:
CsCl
CAS Number:
Molecular Weight:
168.36
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.23

€140.00


Check Cart for Availability

Request a Bulk Order

product line

AnhydroBeads

Quality Level

Assay

99.999% trace metals basis

form

crystals

particle size

-10 mesh

mp

645 °C (lit.)

SMILES string

[Cl-].[Cs+]

InChI

1S/ClH.Cs/h1H;/q;+1/p-1

InChI key

AIYUHDOJVYHVIT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Cesium chloride finds application in synthesis of perovksites based photovoltaic materials[1][2]. Our perovskite grade CsCl can readily be dissolved in DMF to yield 1M solution.
Used for the preparation of electrically conducting glasses.Used to make solutions for the separation of RNA from DNA by density gradient centrifugation.

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qi Li et al.
Advanced materials (Deerfield Beach, Fla.), e1803095-e1803095 (2018-08-25)
The fabrication of high-quality perovskite film highly relies on chemical composition and the synthesis method of perovskite. So far, sequentially deposited MA0.03 FA0.97 Pb(I0.97 Br0.03 )3 polycrystalline film is adopted to produce high-performance perovskite solar cells with record power conversion
Effect of cesium chloride modification on the film morphology and UV-induced stability of planar perovskite solar cells..
Li W, et al.
Journal of Material Chemistry A, 4, 11688-11695 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service