Skip to Content
Merck
All Photos(1)

Key Documents

906271

cis-(−)-Limonene oxide

Synonym(s):

(−)-cis-Limonene 1,2-epoxide, cis-(−)-1,2-Epoxy-p-menth-8-ene, (−)-cis-p-Mentha-1,8-diene 1,2-epoxide, (1S,2R,4S)-4-Isopropenyl-1-methyl-1-cyclohexene 1,2-epoxide, (1S,4S)-1,2-Epoxy-p-menth-8-ene, (1S,4S,6R)-1-Methyl-4-(1-methylethenyl)-7-oxabicyclo[4.1.0]heptane

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€223.00

€223.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€223.00

About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
MDL number:
UNSPSC Code:
12352212
NACRES:
NA.22

€223.00


Check Cart for Availability

Request a Bulk Order

form

liquid

density

0.913 g/mL

functional group

ether

SMILES string

O1[C@@]2([C@H]1C[C@H](CC2)C(=C)C)C

InChI

1S/C10H16O/c1-7(2)8-4-5-10(3)9(6-8)11-10/h8-9H,1,4-6H2,2-3H3/t8-,9+,10-/m0/s1

InChI key

CCEFMUBVSUDRLG-AEJSXWLSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

cis-(−)-Limonene oxide is a valuable chiral terpene oxide building block.

related product

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

163.4 °F

Flash Point(C)

73 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diastereoselective ring opening of limonene oxide with water catalysed by β-ketophosphonate complexes of molybdenum(VI)
Salles L, et al.
Tetrahedron Asymmetry, 10(8), 1471-1476 (1999)
Diastereoselective ring opening of limonene oxide with water catalysed by β-ketophosphonate complexes of molybdenum(VI)
Salles L,et al.
Tetrahedron Asymmetry, 10(8), 1471-1476 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service