Skip to Content
Merck
All Photos(2)

Key Documents

906263

cis-(+)-Limonene oxide

Synonym(s):

(+)-cis-Limonene-1,2-epoxide, (1R,4R,6S)-1-methyl-4-(1-methylethenyl)-7-oxabicyclo[4.1.0]heptane, cis-(+)-1,2-Epoxy-p-menth-8-ene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€397.00
25 G
€796.00

€397.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€397.00
25 G
€796.00

About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
MDL number:
UNSPSC Code:
12352212
NACRES:
NA.22

€397.00


Check Cart for Availability

Request a Bulk Order

form

liquid

density

0.932 g/mL

SMILES string

O1[C@@]2([C@H]1C[C@H](CC2)C(=C)C)C

InChI

1S/C10H16O/c1-7(2)8-4-5-10(3)9(6-8)11-10/h8-9H,1,4-6H2,2-3H3/t8-,9+,10-/m0/s1

InChI key

CCEFMUBVSUDRLG-AEJSXWLSSA-N

Application

cis-(+)-Limonene oxide can be used as:
  • A monomer to prepare poly(limonene)carbonate by Al(III) catalyzed coupling reaction with CO2.[1]
  • A reactant to prepare trans-diaxial diol via diastereoselective ring-opening reaction using molybdenum complex catalyst.[2]
  • A reactant to synthesize trans-dihydrocarvone via isomerization reaction in the presence of silica-supported tungstophosphoric acid as a catalyst and dialkylcarbonate as a green solvent.[3]

cis-(+)-Limonene oxide is a valuable chiral terpene oxide building block.

related product

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

160.7 °F

Flash Point(C)

71.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heteropoly acid catalysis for the isomerization of biomass-derived limonene oxide and kinetic separation of the trans-isomer in green solvents
Cotta RF, et al.
Applied Catalysis A: General, 584, 117173-117173 (2019)
Leticia Peña Carrodeguas et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(16), 6115-6122 (2015-03-05)
Amino-triphenolate derived Al(III) complexes combined with suitable nucleophiles have been investigated as binary catalysts for the coupling of limonene oxide and carbon dioxide to afford alternating polycarbonates. These catalysts are able to produce stereoregular, perfectly alternating trans-polymers from cis-limonene oxide
Diastereoselective ring opening of limonene oxide with water catalysed by β-ketophosphonate complexes of molybdenum(VI)
Salles L,et al.
Tetrahedron Asymmetry, 10(8), 1471-1476 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service