Skip to Content
Merck
All Photos(2)

Key Documents

904805

Sigma-Aldrich

((S)-3-(3,5-Bis(trifluoromethyl)phenyl)-1-(2′-(3-(3,5-bis(trifluoromethyl)phenyl)ureido)-[1,1′-binaphthalen]-2-yl)-1-isopropylurea

≥95%

Synonym(s):

Gouverneur fluorination catalyst

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€287.00

€287.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€287.00

About This Item

Empirical Formula (Hill Notation):
C41H28F12N4O2
CAS Number:
Molecular Weight:
836.67
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

€287.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95%

form

powder or crystals

mp

144-160 °C

SMILES string

FC(F)(F)c1cc(cc(c1)C(F)(F)F)NC(=O)N(C(C)C)c2c(c6c(cc2)cccc6)c3c4c(ccc3NC(=O)Nc5cc(cc(c5)C(F)(F)F)C(F)(F)F)cccc4

InChI

1S/C41H28F12N4O2/c1-21(2)57(37(59)55-29-19-26(40(48,49)50)16-27(20-29)41(51,52)53)33-14-12-23-8-4-6-10-31(23)35(33)34-30-9-5-3-7-22(30)11-13-32(34)56-36(58)54-28-17-24(38(42,43)44)15-25(18-28)39(45,46)47/h3-21H,1-2H3,(H,55,59)(H2,54,56,58)

InChI key

VKRWLTPTRNIRGQ-UHFFFAOYSA-N

Application

((S)-3-(3,5-Bis(trifluoromethyl)phenyl)-1-(2′-(3-(3,5-bis(trifluoromethyl)phenyl)ureido)-[1,1′-binaphthalen]-2-yl)-1-isopropylurea is a hydrogen bonding phase-transfer catalyst capable of activating CsF for the asymmetric nucleophilic fluorination of sulfoniums.1

Automate your fluorination reactions with Synple Automated Synthesis Platform (SYNPLE-SC002)

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriele Pupo et al.
Science (New York, N.Y.), 360(6389), 638-642 (2018-05-12)
Common anionic nucleophiles such as those derived from inorganic salts have not been used for enantioselective catalysis because of their insolubility. Here, we report that merging hydrogen bonding and phase-transfer catalysis provides an effective mode of activation for nucleophiles that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service