Skip to Content
Merck
All Photos(1)

Documents

728357

Sigma-Aldrich

N-[(1R,2R)-2-(1-Piperidinyl)cyclohexyl]-N′-[4-(trifluoromethyl)phenyl]squaramide

95%

Synonym(s):

3-{[(1R,2R)-2-(1-Piperidinyl)cyclohexyl]amino}-4-{[4-(trifluoromethyl)phenyl]amino}-3-cyclobutene-1,2-dione, Rawal Squaramide Organocatalyst

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26F3N3O2
CAS Number:
Molecular Weight:
421.46
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥94.5% (HPLC)
95%

form

powder

optical activity

[α]/D -42.0±2.0°, c = 1 in DMSO

impurities

≤2.0% pyridine

SMILES string

FC(F)(F)c1ccc(NC2=C(N[C@@H]3CCCC[C@H]3N4CCCCC4)C(=O)C2=O)cc1

InChI

1S/C22H26F3N3O2/c23-22(24,25)14-8-10-15(11-9-14)26-18-19(21(30)20(18)29)27-16-6-2-3-7-17(16)28-12-4-1-5-13-28/h8-11,16-17,26-27H,1-7,12-13H2/t16-,17-/m1/s1

InChI key

JRSCPJJNZMRAQS-IAGOWNOFSA-N

Application

Catalyst for enantioselective Michael addition.
It can act as a catalyst for the asymmetric tandem aza-Michael addition-protonation between 1-nitro cyclohexene and 4-methoxyaniline to form the corresponding α-arylamino-β-nitro cyclohexane in a 35:65 trans/cis mixture.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Squaramide-catalyzed enantioselective Michael addition of diphenyl phosphite to nitroalkenes.
Ye Zhu et al.
Angewandte Chemie (International ed. in English), 49(1), 153-156 (2009-12-02)
Stereoselective aza-Michael addition of anilines to 1-nitro cyclohexene by intramolecular protonation.
Ghisu L, et al.
Tetrahedron Letters, 56(46), 6409-6412 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service