Skip to Content
Merck
All Photos(3)

Key Documents

904694

Sigma-Aldrich

Ir(p-CF3-ppy)3

≥95%

Synonym(s):

Tris[2-(2-pyridinyl-κN)-5-(trifluoromethyl)phenyl-κC]iridium

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€177.00
500 MG
€646.00

€177.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€177.00
500 MG
€646.00

About This Item

Empirical Formula (Hill Notation):
C36H21F9IrN3
CAS Number:
Molecular Weight:
858.77
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

€177.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95%

form

powder or crystals

reaction suitability

core: iridium
reagent type: catalyst
reaction type: Photocatalysis

mp

>300 °C

photocatalyst activation

465 nm

SMILES string

[Ir]741([NH]8=C(c9c7cc(cc9)C(F)(F)F)C=CC=C8)([NH]5=C(c6c4cc(cc6)C(F)(F)F)C=CC=C5)[NH]2=C(c3c1cc(cc3)C(F)(F)F)C=CC=C2

InChI

1S/3C12H7F3N.Ir/c3*13-12(14,15)10-6-4-9(5-7-10)11-3-1-2-8-16-11;/h3*1-4,6-8H;

InChI key

ZLVNAIXHGHEODI-UHFFFAOYSA-N

Application

This photocatalyst facilitates a variety of transformations using visible light, including the defluorination of arenes.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Singh et al.
Chemical science, 6(12), 7206-7212 (2015-12-01)
C-F functionalizations that provide C-C bonds are challenging synthetic transformations, due in part to the large C-F bond strength, short bond length, nonpolarizable nature, the production of fluoride, and the regioselectivity-in the case of multifluorinated substrates. However, commercially available highly
Facile synthesis and complete characterization of homoleptic and heteroleptic cyclometalated Iridium(III) complexes for photocatalysis
Singh A, et al.
Journal of Organometallic Chemistry, 776, 51-59 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service