682594
Tris[2-(4,6-difluorophenyl)pyridinato-C2,N]iridium(III)
96%
Synonym(s):
Ir(Fppy)3, Tris[2-(2,4-difluorophenyl)pyridine]iridium(III)
About This Item
Recommended Products
Assay
96%
form
powder
λmax
347 nm
fluorescence
λem 480 nm in chloroform
SMILES string
Fc1cc(F)c(-c2ccccn2)c(c1)[Ir](c3cc(F)cc(F)c3-c4ccccn4)c5cc(F)cc(F)c5-c6ccccn6
InChI
1S/3C11H6F2N.Ir/c3*12-8-4-5-9(10(13)7-8)11-3-1-2-6-14-11;/h3*1-4,6-7H;
InChI key
GFSFWLGQTIKUOK-UHFFFAOYSA-N
Related Categories
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Materials Design Concepts for Efficient Blue OLEDs: A Joint Theoretical and Experimental Study
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service