Skip to Content
Merck
All Photos(2)

Key Documents

904465

Sigma-Aldrich

Silica

monodisperse, non-porous, 2.0 μm

Synonym(s):

Monodisperse silicon dioxide, Non-porous silica, Non-porous silicon dioxide, Silica, Silicon dioxide

Sign Into View Organizational & Contract Pricing

Select a Size

2 G
€327.00

€327.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2 G
€327.00

About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
41123003
NACRES:
NA.23

€327.00


Check Cart for Availability

Request a Bulk Order

form

powder

bp

2230 °C (lit.)

mp

>1600 °C (lit.)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Monodisperse nonporous silica nanoparticles find application in drug delivery and molecular imaging[1][2] whereas micron sized particles are used in liquid chromatography[3]. We offer highly monodisperse(coefficient of variance <10%) nonporous silica in size varying from 150 nm to 50 μm. The high monodispersity of our particles makes them suitable for aforementioned applications.
Silica monodispersed, non-porous (2 μm) is a micro-sized filler material can be used as a spacer in liquid crystals,[4] high performance liquid chromatography (HPLC),[5] and nanomedicine for biological applications.[6]

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Practical Examination of a Nonporous Silica Stationary Phase for Reversed-Phase Fast LC Applications
D R Jenke
Journal of Chromatographic Science, 34, 362-362 (1996)
In-depth characterization of slurry packed capillary columns with 1.0-mm nonporous particles using reversed-phase isocratic ultrahigh-pressure liquid chromatography
Patel KD, et al.
Analytical Chemistry, 76(19), 5777-5786 (2004)
Size control of monodisperse nonporous silica particles by seed particle growth
Zhao B, et al.
Particuology, 9(3), 314-317 (2011)
Nonporous silica nanoparticles for nanomedicine application
Tang L and Cheng J
Nano Today, 8(3), 290-312 (2013)
Martha Kim et al.
Scientific reports, 7(1), 8238-8238 (2017-08-16)
Nonporous silica nanoparticles (SiNPs) have potential as promising carriers for ophthalmic drugs. However, the in vivo safety of ocular topical SiNPs remains unclear. This study investigated the in vivo safety of oral and ocular topical applications of 100 nm-sized SiNPs in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service