Skip to Content
Merck
All Photos(1)

Key Documents

902780

Sigma-Aldrich

Carbon disulfide solution

5.0 M in hexane

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€49.50
100 ML
€117.00

€49.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
€49.50
100 ML
€117.00

About This Item

Empirical Formula (Hill Notation):
CS2
CAS Number:
Molecular Weight:
76.14
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.22

€49.50


Check Cart for Availability

Request a Bulk Order

form

liquid

concentration

5.0 M in hexane

refractive index

n/D 1.434

density

0.832 g/mL

SMILES string

S=C=S

InChI

1S/CS2/c2-1-3

InChI key

QGJOPFRUJISHPQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Valuable building block offered as a solution in hexane for more convenient handling.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system, Peripheral nervous system,Central nervous system,Cardio-vascular system,Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

<-22.0 °F

Flash Point(C)

< -30.0 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service