Skip to Content
Merck
All Photos(1)

Key Documents

115150

Sigma-Aldrich

Thiophosgene

Synonym(s):

Thiocarbonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CSCl2
CAS Number:
Molecular Weight:
114.98
Beilstein:
1633495
EC Number:
MDL number:
UNSPSC Code:
12352000
eCl@ss:
38020402
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

4 (vs air)

Quality Level

reaction suitability

reaction type: Carbonylations

refractive index

n20/D 1.548 (lit.)

bp

70-75 °C (lit.)

density

1.50 g/mL at 20 °C (lit.)

functional group

chloro

storage temp.

2-8°C

SMILES string

ClC(Cl)=S

InChI

1S/CCl2S/c2-1(3)4

InChI key

ZWZVWGITAAIFPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent used to prepare a bulky thiourea ligand for palladium-catalyzed aerobic oxidation of alcohols to aldehydes and ketones.[1]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T A Lewis et al.
Journal of bacteriology, 177(8), 2204-2208 (1995-04-01)
Pseudomonas sp. strain KC transforms carbon tetrachloride into carbon dioxide and nonvolatile products, without chloroform as an intermediate. To define the pathway for hydrolysis, nonvolatile products were analyzed. Condensation products containing the carbon atom of carbon tetrachloride as carbonyl and
Cheng Li et al.
Colloids and surfaces. B, Biointerfaces, 171, 159-166 (2018-07-22)
Dual mode imaging technology is widely developed to achieve the early-stage precision cancer diagnosis. Here we designed a dual-modal magnetic resonance/near infrared fluorescence optical imaging contrast agent (GdF-SS-NIR783) with the fluorescence activatable and safer gadofullerene. The nanoprobes were fabricated by
Christof Jung et al.
The journal of physical chemistry. A, 110(16), 5317-5325 (2006-04-21)
The dispersed fluorescence spectrum of the ground electronic state of thiophosgene, SCCl2, is analyzed in a very complex region of vibrational excitation, 7000-9000 cm(-1). The final result is that most of the inferred excited vibrational levels are assigned in terms
W Kedzierski et al.
The Journal of chemical physics, 120(19), 9087-9089 (2004-07-23)
A special xenon matrix detector has been used to study the production of S(1S) following controlled electron impact on thiophosgene (Cl2CS) targets over an electron energy range from threshold to 400 eV. Time-of-flight spectroscopy has been used to measure S(1S)
Synlett, 3057-3057 (2006)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service