Skip to Content
Merck
All Photos(1)

Documents

901803

Sigma-Aldrich

Fluorescent nanodiamond

nitrogen vacancy ~3 ppm, 140 nm avg. part. size, hydroxyl functionalized, PEG 3000 coated, 1 mg/mL in deionized water

Synonym(s):

Fluorescent nanoparticles, Nanodiamonds

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302
NACRES:
NA.23

form

nanoparticles

composition

nitrogen vacancy, ~3 ppm

concentration

1 mg/mL in deionized water

avg. part. size

140 nm

Looking for similar products? Visit Product Comparison Guide

General description

Fluorescent nanodiamond (nitrogen vacancy ~3 ppm, 140 nm avg. part. size, hydroxyl functionalized, PEG 3000 coated, 1 mg/mL in deionized water), a biocompatible carbon-based nanomaterial, has interesting properties like excellent photostability, low toxic and facile surface functionalizability. It can be used for a variety of biological applications.

Application

Nanodiamonds (NDs) containing NV color centers exhibit fluorescence in red/near infra-red (NIR) region. The fluorescence in NDs originates from nitrogen vacancies and, unlike quantum dots and organic dyes, NDs do not photobleach. Fluorescent NDs find applications in background-free imaging, flow cytometry, super-resolution imaging etc. The PEG coated NDs exhibit reduced non-specific binding and are well suited for biomedical applications.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nanodiamond bioconjugate probes and their collection by electrophoresis.
S. C. Hens et al.,
Diamond and Related Materials, 17, 1858-1858 (2008)
Functionality is Key: Recent Progress in the Surface Modification of Nanodiamond.
A. Krueger a4nd D. Lang
Advances in Functional Materials, 22, 890-890 (2012)
Nanodiamond-mediated drug delivery and imaging: Challenges and opportunities.
V. Vaijayanthimala et al.,
Expert Opinion on Drug Delivery, 12, 1-1 (2014)
Highly Fluorescent Nanodiamonds Protein-Functionalized for Cell Labeling and Targeting
B. Chang et al.,
Advances in Functional Materials, 23, 5737-5737 (2013)
The long-term stability and biocompatibility of fluorescent nanodiamond as an in vivo contrast agent
Vaijayanthimala V, et al.
Biomaterials, 33(31), 7794-7802 (2012)

Articles

Fluorescent Nanodiamond Particles (FNDs)- Find properties and applications of nanodiamond particles.

Fluorescent Nanodiamonds and biofunctionalization strategies are detailed for imaging and bioconjugation applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service