Skip to Content
Merck
All Photos(1)

Key Documents

772372

Sigma-Aldrich

DTS(PTTh2)2

Synonym(s):

4,4′-[4,4-Bis(2-ethylhexyl)-4H-silolo[3,2-b:4,5-b′]dithiophene-2,6-diyl]bis[7-(5′-hexyl-[2,2′-bithiophen]-5-yl)-[1,2,5]thiadiazolo[3,4-c]pyridine], 5,5′-Bis{[4-(7-hexylthiophen-2-yl)thiophen-2-yl]-[1,2,5]thiadiazolo[3,4-c]pyridine}-3,3′-di-2-ethylhexylsilylene-2,2′-bithiophene

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€331.00

€331.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€331.00

About This Item

Empirical Formula (Hill Notation):
C62H72N6S8Si
Molecular Weight:
1185.88
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€331.00


Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

mp

208-213 °C

solubility

chloroform: soluble(lit.)
dichlorobenzene: soluble(lit.)

λmax

655 nm in chloroform

SMILES string

CCC(CCCC)C[Si]1(CC(CCCC)CC)C2=C(SC(C3=NC=C(C4=CC=C(C5=CC=C(CCCCCC)S5)S4)C6=NSN=C63)=C2)C7=C1C=C(C8=NC=C(C9=CC=C(C%10=CC=C(CCCCCC)S%10)S9)C%11=NSN=C%118)S7

InChI

1S/C62H72N6S8Si/c1-7-13-17-19-23-41-25-27-47(69-41)49-31-29-45(71-49)43-35-63-57(59-55(43)65-75-67-59)51-33-53-61(73-51)62-54(77(53,37-39(11-5)21-15-9-3)38-40(12-6)22-16-10-4)34-52(74-62)58-60-56(66-76-68-60)44(36-64-58)46-30-32-50(72-46)48-28-26-42(70-48)24-20-18-14-8-2/h25-36,39-40H,7-24,37-38H2,1-6H3

InChI key

NOJURONZIGXBEP-UHFFFAOYSA-N

General description

DTS(PTTh2)2 is a conjugating polymer with an absorption onset of 815 nm and a field effect hole mobility of ~ 0.1 cm2V-1s-1. It acts as a small donor molecule that can be used as an active layer in optoelectronic applications.[1][2][3][4]

Application

Narrow band gap material for high-efficiency organic solar cells (OPVs) application
OPV Device Structure: ITO/MoOx/DTS(PTTh2)2: PC70BM/Al
  • JSC = 14.4 mA/cm2
  • VOC = 0.78 V
  • FF = 0.59
  • PCE = 6.7%

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zachary B Henson et al.
Journal of the American Chemical Society, 134(8), 3766-3779 (2012-01-31)
π-Conjugated materials containing pyridal[2,1,3]thiadiazole (PT) units have recently achieved record power conversion efficiencies of 6.7% in solution-processed, molecular bulk-heterojunction (BHJ) organic photovoltaics. Recognizing the importance of this new class of molecular systems and with the aim of establishing a more
Optical performance and color investigations of hybrid solar cells based on P3HT: ZnO, PCPDTBT: ZnO, PTB7: ZnO and DTS (PTTh2) 2: ZnO
Duche D, et al.
Solar Energy Materials and Solar Cells, 126, 197-204 (2014)
Plastic solar cells: Self-assembly of bulk heterojunction nano-materials by spontaneous phase separation
AIP Conference Proceedings, 1519(1), 47-50 (2013)
Balancing the H-and J-aggregation in DTS (PTTh 2) 2/PC 70 BM to yield a high photovoltaic efficiency
Zhao Q, et al.
Journal of Material Chemistry C, 3(31), 8183-8192 (2015)
Solution-processed small-molecule solar cells with 6.7% efficiency
Nature Materials, 11, 44-48 (2012)

Articles

Solution-processed organic photovoltaic devices (OPVs) have emerged as a promising clean energy generating technology due to their ease of fabrication, potential to enable low-cost manufacturing via printing or coating techniques, and ability to be incorporated onto light weight, flexible substrates.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service