Skip to Content
Merck
All Photos(1)

Key Documents

791873

Sigma-Aldrich

Conductive silver printing ink, resistivity 5-6 Ω cm

Synonym(s):

Greatcell Solar®, Dyesol(R) DYAG50 conductive silver ink, Screen printable silver paste

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€231.00
20 G
€412.00

€231.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€231.00
20 G
€412.00

About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

€231.00


Check Cart for Availability

Request a Bulk Order

description

volume resistivity 5 - 6 Ω cm

Quality Level

Assay

75-85% solids basis

form

paste (white)

viscosity

13,000-17,000 mPa.s (at shear rate of 10 sec-1 at 25°C)

General description

Sheet Resistivity is 2.0 - 2.4 μΩ/ square / mil. (at a 180°C cure temperature)

Application

Use this Conductive Silver Ink to achieve exceptionally high conductivity at low cure temperatures when printing on a variety of substrates, including glass, polyesters (PET/PEN), and ITO/FTO coated substrates.
This Conductive Silver Printing Ink is a specially formulated ink which provides exceptionally high conductivity at low cure temperatures. This highly conductive ink provides extremely low resistance printed conducting tracks, essential for the photovoltaic applications. By providing the lowest track resistances of printed current collectors, this material is a key enabler in plastic electronics, such as, DSSC, OPV, and CdTe solar cells.

Legal Information

Product of Greatcell Solar Materials Pty Ltd.
GreatcellSolar is a registered trademark of Greatcell Solar Materials Pty Ltd.
Greatcell Solar is a registered trademark of Greatcell Solar

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F

Flash Point(C)

79 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Metin Uz et al.
ACS applied materials & interfaces, 12(11), 13529-13539 (2020-02-23)
In this study, a simple microfluidic method, which can be universally applied to different rigid or flexible substrates, was developed to fabricate high-resolution, conductive, two-dimensional and three-dimensional microstructured graphene-based electronic circuits. The method involves controlled and selective filling of microchannels

Articles

The ability to pattern conductive electrodes is technologically relevant for several applications, including photovolatics, displays, sensors, and biomedical devices.

Small molecular weight organic semiconductors are promising for flexible transistor applications in next-gen soft electronics.

Dye-sensitized solar cells (DSCs) are 3rd generation solar cells combining the promise of high efficiency with low production costs.

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

See All

Questions

1–2 of 2 Questions  
  1. How long does it take to dry naturally after printing ink?

    1 answer
    1. There is no specific information available on the exact drying time after printing ink. Overnight drying is generally recommended, but it is important to consider the substrate and environmental conditions, as these can influence the actual drying time. If precise drying time is required, conducting a test on the specific substrate used may provide better guidance.

      Helpful?

  2. What is the particle size distribution of the colloidal silver in this ink? What would be the best solvent to use to remove the residual ink from a screen printing mesh?

    1 answer
    1. This item is a product of Greatcell Solar Materials. The manufacturer considers the particle size distribution to be proprietary. The preferred solvent for the removal of residual ink is ethanol. However, the end-user should ensure that the screen material is compatible with the ethanol and ink components before use. The ink components can be found in the product Safety Data Sheet.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service