Skip to Content
Merck
All Photos(7)

Documents

901002

Sigma-Aldrich

Carbon nanotube, multi-walled

flake of bundled CNTs, >95% carbon basis, O.D. × L 10-40 nm × 0.5-1.5 mm

Synonym(s):

MWCNT, MWNT, Multiwall carbon nanotube

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352103
NACRES:
NA.23

Quality Level

Assay

>95% carbon basis

O.D. × L

10-40 nm × 0.5-1.5 mm

mp

3652-3697 °C (lit.)

density

~2.1 g/mL at 25 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

General description

This carbon nanotube flakes are millimetre-scale multi-walled carbon nanotube bundles. Crystal quality of CNT is examined by Raman scattering measurement. Intensity ratio of G-peak (1580 cm-1) and D-peak (1350 cm-1) represents quality of graphitization of carbon materials. G/D of the CNT is ∼3, showing high crystal quality.

Preparation Note

Chemical Vapor Deposition (CVD) Method

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service