Skip to Content
Merck
All Photos(3)

Key Documents

755133

Sigma-Aldrich

Carbon nanotube, multi-walled

thin, <5% Metal Oxide(TGA)

Synonym(s):

NC-3100, MWCNT, MWNT, Multiwall carbon nanotube

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€412.00

€412.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€412.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

€412.00


Check Cart for Availability

Request a Bulk Order

Quality Level

form

powder

avg. diam. × L

9.5 nm × 1.5 μm , TEM

impurities

<5% Metal Oxide (TGA)

mp

3652-3697 °C (lit.)

density

~2.1 g/mL at 25 °C (lit.)

SMILES string

[C]

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Multi walled carbon nanotubes (MWNTs, CNTs) were prepared by chemical vapor deposition (CVD). In chemical vapor deposition (CVD), a volatile precursor undergoes thermal decomposition at elevated temperatures to form a solid deposit on a substrate. Compared to other techniques, CVD produces greater yield of carbon nanotubes. Morphology and structure of the nanotubes can be easily tuned using CVD technique. The CNTs formed can be formed in bundles over a catalyst support or in patterns over substrates.1

Application

Applications include:
Electronics[1]
Sensors[2]
Composites
Energy Storage
Study of Life Science system[3]

Preparation Note

Catalytic Carbon Vapor Deposition (CCVD) Method

Legal Information

Product of Nanocyl, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syeda Warisul Fatima et al.
International journal of biological macromolecules, 163, 1747-1758 (2020-09-23)
The chemical cross-linkers are difficult to be removed from the scaffold materials, which limit their application in tissue engineering; designing an efficient biocompatible hydrogel is though challenging is desirable. The aim of the present study was to immobilize microbial Transglutaminase
Dominick J Bindl et al.
ACS nano, 4(10), 5657-5664 (2010-10-07)
Semiconducting single-walled carbon nanotubes (s-SWCNTs) have strong near-infrared and visible absorptivity and exceptional charge transport characteristics, rendering them highly attractive semiconductor absorbers for photovoltaic and photodetector technologies. However, these applications are limited by a poor understanding of how photogenerated charges
Wai Li Ling et al.
ACS nano, 5(2), 730-737 (2011-01-11)
The classical pathway of complement is an essential component of the human innate immune system involved in the defense against pathogens as well as in the clearance of altered self-components. Activation of this pathway is triggered by C1, a multimolecular
Amin Salehi-Khojin et al.
ACS nano, 5(1), 153-158 (2010-12-29)
There has been recent controversy whether the response seen in carbon nanotube (CNT) chemiresistors is associated with a change in the resistance of the individual nanotubes or changes in the resistance of the junctions. In this study, we carry out

Articles

Find unique properties & applications of single (SWNTs) , double (DWNTs) & multi walled carbon nanotubes (MWCNTs).

3D printing is a type of additive manufacturing that can be used to rapidly fabricate components with highly customizable geometries.

Boron nitride nanotubes (BNNT) are close structural analogs of carbon nanotubes (CNT), which are high aspect ratio nanotubular material, where carbon atoms are alternately substituted by nitrogen and boron atoms.

Carbon nanotubes (CNTs) have received much attention since their discovery in 1991 by Sumio lijima1 due to their excellent mechanical, electrical, and optical properties.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service