Skip to Content
Merck
All Photos(2)

Key Documents

900927

Sigma-Aldrich

TCO-PNB ester

≥95%

Synonym(s):

(1R,4E)-4-Cycloocten-1-yl 4-nitrophenyl carbonate, rel-(1R-4E-PS)-cyclooct-4-en-1-yl (4-nitrophenyl) carbonate

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€323.00

€323.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€323.00

About This Item

Empirical Formula (Hill Notation):
C15H17NO5
CAS Number:
Molecular Weight:
291.30
MDL number:
UNSPSC Code:
12352108
NACRES:
NA.22

€323.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

powder or chunks

reaction suitability

reaction type: click chemistry
reagent type: cross-linking reagent

storage temp.

−20°C

SMILES string

O=C(OC1CCC/C=C/CC1)OC2=CC=C([N+]([O-])=O)C=C2

InChI

1S/C15H17NO5/c17-15(20-13-6-4-2-1-3-5-7-13)21-14-10-8-12(9-11-14)16(18)19/h1-2,8-11,13H,3-7H2

InChI key

FJMMADPCDJNXAH-UHFFFAOYSA-N

Application

TCO-PNB ester (trans-Cyclooctene-p-nitrobenzyl ester) has a p-nitrobenzyl (PNB) ester group that is amine reactive and a trans-cyclooctene (TCO) group that is reactive with tetrazines.[1] TCO-PNB ester is supplied as a single axial (major) diastereomer. This specially formulated crystalline material provides easy handling and extended shelf life.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Imaging cell surface glycosylation in vivo using ?double click? chemistry.
Neves A A, et al.
Bioconjugate Chemistry, 24(6), 934-941 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service