Skip to Content
Merck
All Photos(1)

Key Documents

776173

Sigma-Aldrich

5-Norbornene-2-acetic acid succinimidyl ester

97%

Synonym(s):

2,5-Dioxopyrrolidin-1-yl 2-(bicyclo[2.2.1]hept-5-en-2-yl)acetate, 5-Norbornene-2-acetic acid NHS, Bicyclo[2.2.1]hept-5-ene-2-acetic acid N-hydroxysuccinimide ester, Bicyclo[2.2.1]hept-5-ene-2-acetic acid, 2,5-dioxo-1-pyrrolidinyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€130.00
100 MG
€416.00

€130.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€130.00
100 MG
€416.00

About This Item

Empirical Formula (Hill Notation):
C13H15NO4
CAS Number:
Molecular Weight:
249.26
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

€130.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

reaction suitability

reaction type: click chemistry
reagent type: linker

mp

115-120 °C

functional group

NHS ester

storage temp.

2-8°C

SMILES string

O=C(N1OC(C[C@H]2[C@@H]3C=C[C@@H](C3)C2)=O)CCC1=O

InChI

1S/C13H15NO4/c15-11-3-4-12(16)14(11)18-13(17)7-10-6-8-1-2-9(10)5-8/h1-2,8-10H,3-7H2

InChI key

AOGNOQQTUYLDKN-UHFFFAOYSA-N

Application

Succinimidyl ester functionalized norbornene. Succinimidyl ester group will react with amine containing compounds such as lysine residues on proteins/peptides. Norbornene has been shown to react with 1; 2; 4; 5-tetrazines in an inverse electron demand Diels-Alder cycloaddition reaction which is a bioorthogonal reaction demonstrated to be useful in biological imaging applications.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Hee-Sun Han et al.
Journal of the American Chemical Society, 132(23), 7838-7839 (2010-05-21)
We present a bioorthogonal and modular conjugation method for efficient coupling of organic dyes and biomolecules to quantum dots (QDs) using a norbornene-tetrazine cycloaddition. The use of noncoordinating functional groups combined with the rapid rate of the cycloaddition leads to
Neal K Devaraj et al.
Bioconjugate chemistry, 19(12), 2297-2299 (2008-12-05)
Bioorthogonal tetrazine cycloadditions have been applied to live cell labeling. Tetrazines react irreversibly with the strained dienophile norbornene forming dihydropyrazine products and dinitrogen. The reaction is high yielding, selective, and fast in aqueous media. Her2/neu receptors on live human breast

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service