Skip to Content
Merck
All Photos(1)

Key Documents

900875

Sigma-Aldrich

2,2′-[[(1,2-Diphenyl-1,2-ethenediyl]di-4,1-phenylene]bis[4,4,5,5-tetramethyl-1,3,2-dioxaborolane]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H42B2O4
CAS Number:
Molecular Weight:
584.36
UNSPSC Code:
12352103

form

powder

storage temp.

2-8°C

General description

AIE (aggregation-induced emission)-active building block.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A self-assembly induced emission system constructed by the host-guest interaction of AIE-active building blocks.
Bai W, et al.
Chemical Communications (Cambridge, England), 51(6), 1089-1091 (2015)
A highly sensitive and selective fluorescent probe for hypochlorite in pure water with aggregation induced emission characteristics.
Wang C, et al.
Faraday Discussions, 196, 427-438 (2017)
Fluorescent Porous Organic Frameworks Containing Molecular Rotors for Size-Selective Recognition.
Dong J, et al.
Chemistry of Materials, 28(21), 7889-7897 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service