Skip to Content
Merck
All Photos(1)

Key Documents

900770

Sigma-Aldrich

Berkessel ligand

98%

Synonym(s):

2′,3′,4′,5′,6′-Pentafluoro-3-({(E)-[(1R,2S)-2-{[(2′,3′,4′,5′,6′-pentafluoro-2-hydroxy[1,1′-biphenyl]-3-yl)methyl]amino}cyclohexyl]imino}methyl)[1,1′-biphenyl]-2-ol, cis-1,2-Diaminocyclohexane salalen ligand, cis-DACH salalen ligand, Bis(pentafluorophenyl) salalen ligand

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€154.00

€154.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€154.00

About This Item

Empirical Formula (Hill Notation):
C32H22F10N2O2
CAS Number:
Molecular Weight:
656.51
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

€154.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

powder or crystals

reaction suitability

reagent type: ligand

SMILES string

Fc1c(c(c(c(c1F)F)c2c(c(ccc2)CN[C@@H]3[C@@H](CCCC3)\N=C\c4c(c(ccc4)c5c(c(c(c(c5F)F)F)F)F)O)O)F)F

InChI

1S/C32H22F10N2O2/c33-21-19(22(34)26(38)29(41)25(21)37)15-7-3-5-13(31(15)45)11-43-17-9-1-2-10-18(17)44-12-14-6-4-8-16(32(14)46)20-23(35)27(39)30(42)28(40)24(20)36/h3-8,11,17-18,44-46H,1-2,9-10,12H2/b43-11+/t17-,18+/m1/s1

InChI key

NLJCGFOPEQFWQJ-XOZUKITKSA-N

Application

Developed by Albrecht Berkessel′s lab, this is the first catalyst system that allows catalytic enantioselective epoxidation of non-conjugated terminal double bonds with hydrogen peroxide as the oxidant. Eradicating multiple steps and kinetic resolution, this chiral ligand, when used with Ti(OiPr)4 provides a one-step route to enantiopure epoxides that are important intermediates for synthesis of bioactive compounds. Catalyst is recyclable under some conditions. For best results, pentafluorobenzoic acid (P5360) or tetra-n-butylammonium hydrogensulfate (155837) is recommended as a co-catalyst.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service