Skip to Content
Merck
All Photos(1)

Key Documents

803839

Sigma-Aldrich

Graphene dispersion

10 mg/mL,dispersion in NMP

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€301.00

€301.00


Estimated to ship on31 March 2025


Request a Bulk Order

Select a Size

Change View
5 ML
€301.00

About This Item

UNSPSC Code:
12141908
NACRES:
NA.23

€301.00


Estimated to ship on31 March 2025


Request a Bulk Order

Product Name

Graphene dispersion, 10 mg/mL, dispersion in NMP

Quality Level

form

dispersion (in NMP)

composition

Graphene, 1 wt. %
NMP, 99 wt. %

concentration

10 mg/mL

sheet resistance

<103 Ω/sq (graphene)

thickness

1-10 nm , graphene

storage temp.

2-8°C

General description

The conductive graphene sheets can also be re-dispersed in other organic solvents such as ethanol, acetone, toluene and chloroform.
No any surfactants or stabilizers exist on the graphene surface.

Application

  • Graphene-based composites
  • Sensors
  • Biomedicines
  • Nanodevices

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

195.8 °F

Flash Point(C)

91 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Graphene is a unique two-dimensional (2D) structure of monolayer carbon atoms packed into a dense honeycomb crystal that has attracted great interest due to its diverse and fascinating properties.

Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.

Catalytic water splitting produces hydrogen crucial for renewable energy, petroleum refining, and chemical industry applications like methanol production.

Advances in scalable synthesis and processing of two-dimensional materials

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service