Skip to Content
Merck
All Photos(2)

Key Documents

900674

Sigma-Aldrich

Triaminetetraacetate, sodium salt-functionalized silica gel

≥99%, molecular loading ≥0.41 mmol/g

Synonym(s):

Si-TAAcONa, SiliaMetS® TAAcONa

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€93.00

€93.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€93.00

About This Item

UNSPSC Code:
12352116
NACRES:
NA.22

€93.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99%

form

powder

shelf life

2 yr (expiration date period)

reaction suitability

reagent type: catalyst
reagent type: chelator

pore size

60 Å pore size

capacity

0.41 mmol/g loading(molecular loading)

solubility

all common solvents: soluble (aqueous and organic)

bulk density

635 g/L (typical tap density)

Application

Triaminetetraacetate, sodium salt-functionalized silica gel is a metal scavenger: Ca, Cu, Cs, Ir, Li, Mg, Ni, Os, Pd, and Sn.

Caution

For prolonged storage, recommended conditions are to store product in a cool place (<8 °C) under inert atmosphere (argon) when possible.

Legal Information

SiliaMetS is a registered trademark of Silicycle, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service