Skip to Content
Merck
All Photos(1)

Key Documents

900574

Sigma-Aldrich

(S)-2-Phenylpiperidine

95%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€449.00

€449.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€449.00

About This Item

Empirical Formula (Hill Notation):
C11H15N
CAS Number:
Molecular Weight:
161.24
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

€449.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

liquid

functional group

phenyl

storage temp.

2-8°C

SMILES string

N1[C@@H](CCCC1)c2ccccc2

InChI

1S/C11H15N/c1-2-6-10(7-3-1)11-8-4-5-9-12-11/h1-3,6-7,11-12H,4-5,8-9H2/t11-/m0/s1

InChI key

WGIAUTGOUJDVEI-NSHDSACASA-N

Application

Chiral building block developed using Liverpool ChiroChem-patented technology.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service