Skip to Content
Merck
All Photos(1)

Key Documents

P7764

Sigma-Aldrich

L-Proline t-butyl ester

≥98%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€108.00

€108.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€108.00

About This Item

Empirical Formula (Hill Notation):
C9H17NO2
CAS Number:
Molecular Weight:
171.24
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

€108.00


Check Cart for Availability

Request a Bulk Order

Product Name

L-Proline t-butyl ester,

Assay

≥98%

form

liquid

color

clear colorless to faint yellow

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)[C@@H]1CCCN1

InChI

1S/C9H17NO2/c1-9(2,3)12-8(11)7-5-4-6-10-7/h7,10H,4-6H2,1-3H3/t7-/m0/s1

InChI key

XJJBXZIKXFOMLP-ZETCQYMHSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service