Skip to Content
Merck
All Photos(1)

Key Documents

900571

Sigma-Aldrich

Poly(D,L-lactide-co-glycolide) ester terminated

lactide:glycolide 80:20, Mw 200,000

Synonym(s):

PLGA 85:15

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€408.00

€408.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€408.00

About This Item

Linear Formula:
[C3H4O2]x[C2H2O2]y
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

€408.00


Check Cart for Availability

Request a Bulk Order

form

crystals

Quality Level

feed ratio

lactide:glycolide 80:20

mol wt

Mw 200,000

degradation timeframe

<9 months

storage temp.

2-8°C

SMILES string

O2C(C(=O)OC(C2=O)C)C.O1CC(=O)OCC1=O

InChI

1S/C6H8O4.C4H4O4/c1-3-5(7)10-4(2)6(8)9-3;5-3-1-7-4(6)2-8-3/h3-4H,1-2H3;1-2H2

InChI key

LCSKNASZPVZHEG-UHFFFAOYSA-N

Application

Controlled release, nanoparticles, scaffolds.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service